Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Urgcp
Entrez Gene ID
305493
Gene Name
upregulator of cell proliferation
Gene Aliases
RGD1564681, Urg4
Chromosome Location
Chr.14: 85957726 - 86002219 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077660.2 NP_001071128.1 4 - 5 193 70
XM_006251312.2 XP_006251374.1 4 - 5 654 70
XM_006251313.2 XP_006251375.1 4 - 5 483 70
XM_006251315.2 XP_006251377.1 4 - 5 286 70
GenBank mRNA CB720935.1 - 2 - 3 71 70
CB757259.1 - 4 - 5 219 70
CB809919.1 - 4 - 5 187 70
FM044330.1 - 4 - 5 193 70
FM046879.1 - 3 - 4 143 70
FQ209875.1 - 4 - 5 213 70
FQ222248.1 - 4 - 5 185 70
FQ223767.1 - 4 - 5 180 70

View Full Product Details