The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024142.1 | NP_077056.1 | 2 - 3 | 114 | 68 |
XM_006250634.2 | XP_006250696.1 | 1 - 2 | 182 | 68 | |
GenBank mRNA | AF260922.1 | - | 2 - 3 | 114 | 68 |
AF530558.1 | - | 2 - 3 | 111 | 68 |