Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gfra1
Entrez Gene ID
25454
Gene Name
GDNF family receptor alpha 1
Gene Aliases
-
Chromosome Location
Chr.1: 279205514 - 279572976 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012959.1 NP_037091.1 3 - 4 667 97
XM_008760512.1 XP_008758734.1 4 - 5 810 97
XM_008760513.1 XP_008758735.1 4 - 5 674 97
GenBank mRNA U59486.1 - 3 - 4 712 97
U97142.1 - 3 - 4 667 97

View Full Product Details