Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dnmbp
Entrez Gene ID
309362
Gene Name
dynamin binding protein
Gene Aliases
-
Chromosome Location
Chr.1: 263625503 - 263718782 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006231570.2 XP_006231632.1 12 - 13 3403 94
XM_008760486.1 XP_008758708.1 12 - 13 3912 94

View Full Product Details