The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031622.2 | NP_113810.2 | 5 - 6 | 1829 | 77 |
XM_006243409.2 | XP_006243471.1 | 5 - 6 | 2323 | 77 | |
GenBank mRNA | BC109380.1 | - | 5 - 6 | 1780 | 77 |
M64301.1 | - | 4 - 5 | 1457 | 77 |