Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pcbp4
Entrez Gene ID
363133
Gene Name
poly(rC) binding protein 4
Gene Aliases
-
Chromosome Location
Chr.8: 115154720 - 115165706 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191883.1 NP_001178812.1 10 - 11 919 112
XM_006243802.2 XP_006243864.1 11 - 12 1415 112
XM_006243803.2 XP_006243865.1 11 - 12 1414 112

View Full Product Details