Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Acvr2b
Entrez Gene ID
25366
Gene Name
activin A receptor, type IIB
Gene Aliases
-
Chromosome Location
Chr.8: 128087315 - 128118844 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031554.1 NP_113742.1 10 - 11 1561 74
XM_006244072.2 XP_006244134.1 9 - 10 1663 74
GenBank mRNA BC097358.1 - 10 - 11 1561 74
L10640.1 - 8 - 9 1005 74
M87067.1 - 10 - 11 1803 74

View Full Product Details