Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Acvr2b
Entrez Gene ID
25366
Gene Name
activin A receptor, type IIB
Gene Aliases
-
Chromosome Location
Chr.8: 128087315 - 128118844 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031554.1 NP_113742.1 3 - 4 586 106
XM_006244072.2 XP_006244134.1 3 - 4 616 106
GenBank mRNA BC097358.1 - 3 - 4 586 106
M87067.1 - 3 - 4 823 106

View Full Product Details