Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tgm4
Entrez Gene ID
64679
Gene Name
transglutaminase 4
Gene Aliases
Dp1
Chromosome Location
Chr.8: 132117273 - 132157485 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022713.2 NP_073204.2 8 - 9 998 60
XM_006244202.2 XP_006244264.1 8 - 9 1178 60
GenBank mRNA BC066665.1 - 8 - 9 987 60

View Full Product Details