Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Amt
Entrez Gene ID
306586
Gene Name
aminomethyltransferase
Gene Aliases
-
Chromosome Location
Chr.8: 117068388 - 117078633 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014004.1 NP_001014026.1 5 - 6 890 73
GenBank mRNA BC083803.1 - 5 - 6 890 73

View Full Product Details