Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc36
Entrez Gene ID
680018
Gene Name
coiled-coil domain containing 36
Gene Aliases
-
Chromosome Location
Chr.8: 117183274 - 117215398 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001055346.5 XP_001055346.3 3 - 4 308 94

View Full Product Details