The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012640.2 | NP_036772.2 | 3 - 4 | 410 | 89 |
XM_006243607.1 | XP_006243669.1 | 4 - 5 | 590 | 89 | |
GenBank mRNA | CN542437.1 | - | 1 - 2 | 89 | |
M13949.1 | - | 3 - 4 | 410 | 89 |