Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bmp5
Entrez Gene ID
315824
Gene Name
bone morphogenetic protein 5
Gene Aliases
-
Chromosome Location
Chr.8: 82669466 - 82950273 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108168.1 NP_001101638.1 1 - 2 515 132

View Full Product Details