Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mmp13
Entrez Gene ID
171052
Gene Name
matrix metallopeptidase 13
Gene Aliases
-
Chromosome Location
Chr.8: 5522739 - 5533018 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133530.1 NP_598214.1 8 - 9 1223 98
GenBank mRNA M60616.1 - 8 - 9 1196 98

View Full Product Details