Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gbp5
Entrez Gene ID
362050
Gene Name
guanylate binding protein 5
Gene Aliases
-
Chromosome Location
Chr.2: 248178441 - 248196873 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108569.3 NP_001102039.2 2 - 3 429 83
GenBank mRNA FM048046.1 - 2 - 3 429 83

View Full Product Details