Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nr4a3
Entrez Gene ID
58853
Gene Name
nuclear receptor subfamily 4, group A, member 3
Gene Aliases
NOR-2
Chromosome Location
Chr.5: 63781801 - 63822890 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008763702.1 XP_008761924.1 2 - 3 2422 72

View Full Product Details