Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kif27
Entrez Gene ID
246209
Gene Name
kinesin family member 27
Gene Aliases
Krp5
Chromosome Location
Chr.17: 6701285 - 6776450 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198050.1 NP_932167.1 15 - 16 3504 113
XM_006253556.2 XP_006253618.1 15 - 16 3648 113
XM_006253557.2 XP_006253619.1 14 - 15 3504 113
XM_008771424.1 XP_008769646.1 15 - 16 3672 113

View Full Product Details