Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Potea
Entrez Gene ID
305491
Gene Name
POTE ankyrin domain family, member A
Gene Aliases
Ankrd36, Ankrd48, Spergen2
Chromosome Location
Chr.14: 85814809 - 85930536 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008770390.1 XP_008768612.1 10 - 11 1286 72
XM_008770391.1 XP_008768613.1 10 - 11 1286 72
XM_008770392.1 XP_008768614.1 9 - 10 1112 72
XM_008770393.1 XP_008768615.1 5 - 6 2028 72
GenBank mRNA AB095021.1 - 10 - 11 1155 72
BC083629.1 - 10 - 11 1260 72

View Full Product Details