Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Potea
Entrez Gene ID
305491
Gene Name
POTE ankyrin domain family, member A
Gene Aliases
Ankrd36, Ankrd48, Spergen2
Chromosome Location
Chr.14: 85814809 - 85930536 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008770390.1 XP_008768612.1 3 - 4 731 91
XM_008770391.1 XP_008768613.1 3 - 4 731 91
GenBank mRNA AB095021.1 - 3 - 4 600 91
BC083629.1 - 3 - 4 705 91

View Full Product Details