Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hsd17b13
Entrez Gene ID
305150
Gene Name
hydroxysteroid (17-beta) dehydrogenase 13
Gene Aliases
-
Chromosome Location
Chr.14: 7113544 - 7128218 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009684.1 NP_001009684.1 1 - 2 280 60
GenBank mRNA BC088191.1 - 1 - 2 280 60

View Full Product Details