Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam35a
Entrez Gene ID
364514
Gene Name
family with sequence similarity 35, member A
Gene Aliases
RGD1310009
Chromosome Location
Chr.16: 10570307 - 10661528 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025028.2 NP_001020199.2 9 - 10 2708 97
XM_008771135.1 XP_008769357.1 8 - 9 2638 97
XM_008771136.1 XP_008769358.1 7 - 8 2442 97
XM_008771137.1 XP_008769359.1 7 - 8 2406 97
GenBank mRNA FM098539.1 - 2 - 3 357 97

View Full Product Details