Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vstm4
Entrez Gene ID
361112
Gene Name
V-set and transmembrane domain containing 4
Gene Aliases
RGD1310423
Chromosome Location
Chr.16: 9092307 - 9172855 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029920.1 NP_001025091.1 6 - 7 798 94
GenBank mRNA BC098063.1 - 6 - 7 798 94

View Full Product Details