Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Spcs1
Entrez Gene ID
290555
Gene Name
signal peptidase complex subunit 1 homolog (S. cerevisiae)
Gene Aliases
RGD1309125
Chromosome Location
Chr.16: 7080389 - 7081957 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001131006.1 NP_001124478.1 2 - 3 229 88
GenBank mRNA CV795143.1 - 2 - 3 88

View Full Product Details