Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dek
Entrez Gene ID
306817
Gene Name
DEK proto-oncogene
Gene Aliases
-
Chromosome Location
Chr.17: 17965858 - 17987836 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004255.1 NP_001004255.1 3 - 4 357 146
XM_006253756.2 XP_006253818.1 3 - 4 380 146
GenBank mRNA BC079344.1 - 3 - 4 357 146

View Full Product Details