Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Taldo1
Entrez Gene ID
83688
Gene Name
transaldolase 1
Gene Aliases
-
Chromosome Location
Chr.1: 214375555 - 214385886 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031811.3 NP_113999.2 4 - 5 481 61
GenBank mRNA BC059126.1 - 4 - 5 485 61
FQ213939.1 - 4 - 5 493 61
FQ219168.1 - 4 - 5 507 61

View Full Product Details