Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dmrt2
Entrez Gene ID
309430
Gene Name
doublesex and mab-3 related transcription factor 2
Gene Aliases
-
Chromosome Location
Chr.1: 243662819 - 243669312 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107597.1 NP_001101067.1 1 - 2 343 85
XM_008760306.1 XP_008758528.1 1 - 2 347 85
XM_008760307.1 XP_008758529.1 1 - 2 347 85

View Full Product Details