Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zic3
Entrez Gene ID
367944
Gene Name
Zic family member 3
Gene Aliases
RGD1561261
Chromosome Location
Chr.X: 140875468 - 140888344 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109019.2 NP_001102489.1 2 - 3 1144 139

View Full Product Details