Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Krba1
Entrez Gene ID
362358
Gene Name
KRAB-A domain containing 1
Gene Aliases
RGD1306967
Chromosome Location
Chr.4: 78023502 - 78046335 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006236447.2 XP_006236509.1 11 - 12 3233 106
XM_006236448.2 XP_006236510.1 11 - 12 3246 106
XM_008762931.1 XP_008761153.1 11 - 12 3230 106
XM_342681.5 XP_342682.2 11 - 12 3215 106

View Full Product Details