Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fhl2
Entrez Gene ID
63839
Gene Name
four and a half LIM domains 2
Gene Aliases
-
Chromosome Location
Chr.9: 49920611 - 49964902 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006244849.2 XP_006244911.1 1 - 2 2114 134

View Full Product Details