Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tkt
Entrez Gene ID
64524
Gene Name
transketolase
Gene Aliases
-
Chromosome Location
Chr.16: 6609670 - 6634608 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022592.1 NP_072114.1 3 - 4 335 94
XM_006252661.1 XP_006252723.1 2 - 3 367 94
GenBank mRNA U09256.1 - 3 - 4 335 94

View Full Product Details