Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Glud1
Entrez Gene ID
24399
Gene Name
glutamate dehydrogenase 1
Gene Aliases
Ac2-281, Gdh1, Glud, Gludeha, MRG-2
Chromosome Location
Chr.16: 10661911 - 10695557 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012570.2 NP_036702.1 8 - 9 1369 58
GenBank mRNA BC081841.1 - 8 - 9 1253 58
X14044.1 - 8 - 9 1259 58
X14223.1 - 8 - 9 1232 58

View Full Product Details