Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cyp3a18
Entrez Gene ID
252931
Gene Name
cytochrome P450, family 3, subfamily a, polypeptide 18
Gene Aliases
-
Chromosome Location
Chr.12: 10636294 - 10684273 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145782.1 NP_665725.1 10 - 11 1088 92
GenBank mRNA D38381.1 - 10 - 11 1088 92
X79991.1 - 10 - 11 1088 92

View Full Product Details