Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wdr74
Entrez Gene ID
690229
Gene Name
WD repeat domain 74
Gene Aliases
-
Chromosome Location
Chr.1: 224933913 - 224938659 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109569.1 NP_001103039.1 1 - 2 102 72
XM_006230999.2 XP_006231061.1 1 - 2 109 72
XM_006231000.2 XP_006231062.1 1 - 2 109 72
GenBank mRNA BC158834.1 - 1 - 2 71 72

View Full Product Details