Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Aldh3b2
Entrez Gene ID
688800
Gene Name
aldehyde dehydrogenase 3 family, member B2
Gene Aliases
-
Chromosome Location
Chr.1: 219215023 - 219218488 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008760187.1 XP_008758409.1 - 746 104

View Full Product Details