Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dfnb31
Entrez Gene ID
313255
Gene Name
deafness, autosomal recessive 31
Gene Aliases
Cip98, Whrn
Chromosome Location
Chr.5: 79235541 - 79317206 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181088.1 NP_851602.1 9 - 10 1725 82
GenBank mRNA AY227205.1 - 9 - 10 1725 82

View Full Product Details