Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Smoc2
Entrez Gene ID
292401
Gene Name
SPARC related modular calcium binding 2
Gene Aliases
-
Chromosome Location
Chr.1: 56242289 - 56374106 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106215.2 NP_001099685.1 2 - 3 427 72
XM_006227958.2 XP_006228020.1 2 - 3 484 72
GenBank mRNA BC168953.1 - 2 - 3 427 72

View Full Product Details