Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pon1
Entrez Gene ID
84024
Gene Name
paraoxonase 1
Gene Aliases
-
Chromosome Location
Chr.4: 30249749 - 30276297 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032077.1 NP_114466.1 5 - 6 512 65
GenBank mRNA BC091403.1 - 5 - 6 553 65
CO570909.1 - 5 - 6 512 65
FQ209439.1 - 5 - 6 530 65
FQ209632.1 - 2 - 3 208 65
FQ211027.1 - 5 - 6 523 65
FQ211041.1 - 5 - 6 566 65
FQ218360.1 - 5 - 6 527 65
FQ218888.1 - 5 - 6 523 65
FQ219478.1 - 5 - 6 515 65
FQ219506.1 - 5 - 6 523 65
FQ219750.1 - 5 - 6 517 65
FQ219776.1 - 5 - 6 523 65
FQ223824.1 - 4 - 5 423 65
U94856.1 - 5 - 6 470 65

View Full Product Details