Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pon2
Entrez Gene ID
296851
Gene Name
paraoxonase 2
Gene Aliases
-
Chromosome Location
Chr.4: 30344705 - 30380232 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013082.1 NP_001013100.1 3 - 4 255 62
XM_008762699.1 XP_008760921.1 3 - 4 368 62
GenBank mRNA BC079462.1 - 3 - 4 255 62

View Full Product Details