The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013082.1 | NP_001013100.1 | 6 - 7 | 748 | 131 |
XM_008762699.1 | XP_008760921.1 | 6 - 7 | 825 | 131 | |
GenBank mRNA | BC079462.1 | - | 6 - 7 | 748 | 131 |