Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zc3hav1l
Entrez Gene ID
362341
Gene Name
zinc finger CCCH-type, antiviral 1-like
Gene Aliases
RGD1309236
Chromosome Location
Chr.4: 65988820 - 66003450 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_342661.6 XP_342662.5 3 - 4 1782 70

View Full Product Details