Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Irf1
Entrez Gene ID
24508
Gene Name
interferon regulatory factor 1
Gene Aliases
-
Chromosome Location
Chr.10: 39109530 - 39116532 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012591.1 NP_036723.1 5 - 6 746 85
GenBank mRNA BC076382.1 - 5 - 6 659 85
M34253.1 - 5 - 6 746 85

View Full Product Details