Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lyar
Entrez Gene ID
289707
Gene Name
Ly1 antibody reactive
Gene Aliases
-
Chromosome Location
Chr.14: 77308211 - 77321931 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011911.1 NP_001011911.1 4 - 5 497 70
XM_006251087.1 XP_006251149.1 4 - 5 503 70
GenBank mRNA BC079008.1 - 4 - 5 497 70

View Full Product Details