The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113570.1 | NP_001107042.1 | 1 - 2 | 354 | 82 |
GenBank mRNA | CB616240.1 | - | 1 - 2 | 362 | 82 |