The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022231.2 | NP_071567.1 | 7 - 8 | 1620 | 63 |
XM_006257506.2 | XP_006257568.1 | 6 - 7 | 1446 | 63 | |
XM_006257508.2 | XP_006257570.1 | 6 - 7 | 1560 | 63 | |
XM_006257509.2 | XP_006257571.1 | 6 - 7 | 1461 | 63 | |
XM_008773529.1 | XP_008771751.1 | 7 - 8 | 1587 | 63 | |
GenBank mRNA | AB033366.1 | - | 7 - 8 | 1619 | 63 |
AF183429.1 | - | 5 - 6 | 1290 | 63 | |
AF304334.1 | - | - | 2215 | 63 |