Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hs6st2
Entrez Gene ID
302489
Gene Name
heparan sulfate 6-O-sulfotransferase 2
Gene Aliases
RGD1563872, RGD1564397
Chromosome Location
Chr.X: 138675326 - 138972781 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191726.1 NP_001178655.1 3 - 4 979 93

View Full Product Details