The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022400.1 | NP_071795.1 | 10 - 11 | 1149 | 77 |
XM_006229149.1 | XP_006229211.1 | 10 - 11 | 1326 | 77 | |
XM_008759413.1 | XP_008757635.1 | 10 - 11 | 1876 | 77 | |
GenBank mRNA | BC061790.1 | - | 10 - 11 | 1130 | 77 |
U68417.1 | - | 10 - 11 | 1149 | 77 |