Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zfp37
Entrez Gene ID
115768
Gene Name
zinc finger protein 37
Gene Aliases
-
Chromosome Location
Chr.5: 77938261 - 77945322 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058209.2 NP_478116.2 - 334 87
GenBank mRNA AF072439.1 - - 359 87

View Full Product Details