Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gatb
Entrez Gene ID
361974
Gene Name
glutamyl-tRNA(Gln) amidotransferase, subunit B
Gene Aliases
Pet112l
Chromosome Location
Chr.2: 184600693 - 184679980 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003749312.3 XP_003749360.1 3 - 4 487 69

View Full Product Details