Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ptms
Entrez Gene ID
83801
Gene Name
parathymosin
Gene Aliases
ZnBP
Chromosome Location
Chr.4: 157435371 - 157439507 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031975.2 NP_114181.1 2 - 3 445 60
GenBank mRNA BC167753.1 - 2 - 3 435 60
M20616.1 - 1 - 2 49 60
M33025.1 - 1 - 2 49 60
X16481.1 - 2 - 3 228 60

View Full Product Details