Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Akap3
Entrez Gene ID
312720
Gene Name
A kinase (PRKA) anchor protein 3
Gene Aliases
-
Chromosome Location
Chr.4: 159399686 - 159425512 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006237489.2 XP_006237551.1 2 - 3 272 102

View Full Product Details